Learn More
Medchemexpress LLC Amoxicillin | 26787-78-0 | ≥98.0% | 50 G
SDP
Catalog No. 5000406432
Encompass_Preferred
Amoxicillin, also known as Amoxycillin, is an antibiotic characterized by its good oral absorption and broad-spectrum antimicrobial activity. It operates by inhibiting the biosynthesis of polypeptides crucial for bacterial cell wall formation, which in turn effectively suppresses bacterial cell growth.
  • Good oral absorption
  • Broad-spectrum antimicrobial activity
  • Inhibits biosynthesis of bacterial cell wall polypeptides
  • Effective in suppressing bacterial cell growth
Catalog No. 50-004-06432 Supplier Medchemexpress LLC Supplier No. HYB0467A50G
Amoxicillin, also known as Amoxycillin, is an antibiotic characterized by its good oral absorption and broad-spectrum antimicrobial activity. It operates by inhibiting the biosynthesis of polypeptides crucial for bacterial cell wall formation, which in turn effectively suppresses bacterial cell growth.
  • Good oral absorption
  • Broad-spectrum antimicrobial activity
  • Inhibits biosynthesis of bacterial cell wall polypeptides
  • Effective in suppressing bacterial cell growth