Learn More
Medchemexpress LLC Amoxicillin | 26787-78-0 | 99.9% | 10 G
SDP
Catalog No. 5000407803
Encompass_Preferred
Amoxicillin is an antibiotic known for its good oral absorption and broad-spectrum antimicrobial activity. It functions by inhibiting the biosynthesis of polypeptides in the bacterial cell wall, which in turn hinders cell growth.
  • Possesses good oral absorption
  • Provides broad-spectrum antimicrobial activity
  • Inhibits bacterial cell wall biosynthesis
  • Used in various cellular and in vivo research models
Catalog No. 50-004-07803 Supplier Medchemexpress LLC Supplier No. HYB0467A10G
Amoxicillin is an antibiotic known for its good oral absorption and broad-spectrum antimicrobial activity. It functions by inhibiting the biosynthesis of polypeptides in the bacterial cell wall, which in turn hinders cell growth.
  • Possesses good oral absorption
  • Provides broad-spectrum antimicrobial activity
  • Inhibits bacterial cell wall biosynthesis
  • Used in various cellular and in vivo research models