Learn More
Med Vet International Amoxicillin Capsules (Amoxil), 500mg, 500ct
Catalog No. 502827843
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Amoxicillin is a generic version of Amoxil. Amoxicillin is a penicillin-type antibiotic and should be stored at room temperature and way from light and moisture.
Catalog No. 50-282-7843 Supplier Med Vet International Supplier No. RX65862001705
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Amoxicillin is a generic version of Amoxil. Amoxicillin is a penicillin-type antibiotic and should be stored at room temperature and way from light and moisture.