Learn More
Selleck Chemical LLC Amoxicillin S3015-50mg
SDP
Catalog No. 503108240
Encompass_Preferred
Amoxicillin (Amoxycillin) is a moderate-spectrum bacteriolytic -lactam antibiotic used to treat bacterial infections caused by susceptible microorganisms
Catalog No. 50-310-8240 Supplier Selleck Chemical LLC Supplier No. S301550MG
Amoxicillin (Amoxycillin) is a moderate-spectrum bacteriolytic -lactam antibiotic used to treat bacterial infections caused by susceptible microorganisms