Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Amoxicillin, 96%

Catalog No. AC455140250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455140250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC455140250 Supplier Thermo Scientific Chemicals Supplier No. 455140250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 26787-78-0 | C16H19N3O5S | 365.404 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26787-78-0
Molecular Formula C16H19N3O5S
Molecular Weight (g/mol) 365.404
MDL Number MFCD00056860
InChI Key LSQZJLSUYDQPKJ-NJBDSQKTSA-N
PubChem CID 33613
ChEBI CHEBI:2676
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)O)C

Specifications

Type Amoxicillin
Physical Form Solid
Melting Point 330.0°C
Odor Odorless
Quantity 25 g
Formula Weight 365.4
Percent Purity 96%
Concentration or Composition (by Analyte or Components) 4-Thia-1-azobicyclo[3.2.0]heptane-2-carboxylic acid,6-[[amino(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-,[2S-[2.alpha.,5.alpha.,6.beta.(S)]]-
Chemical Name or Material Amoxicillin
Hazard Category Danger
Hazard Statement H317+H334: May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled
Precautionary Statement P280+P304 + P341+P342 + P311+P302 + P352+ P333 + P313: Wear protective gloves/protective clothing.IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.IF ON SKIN: Wash with plenty of soap and water.If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.