Learn More
MOLECULAR DEPOT LLC Amoxicillin BSA - 1 mg
Catalog No. 5000013458
Encompass_Preferred
Amoxicillin BSA conjugate consists of amoxicillin conjugated to bovine serum albumin (BSA) This conjugation enhances immunogenicity aiding in antibody creation for research This product has been used as a molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only
Catalog No. 50-000-13458 Supplier MOLECULAR DEPOT LLC Supplier No. B2020849
Amoxicillin BSA conjugate consists of amoxicillin conjugated to bovine serum albumin (BSA) This conjugation enhances immunogenicity aiding in antibody creation for research This product has been used as a molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only