Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amoxicillin Crystalline MP Biomedicals

Catalog No. ICN19014525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19014525 25 g
ICN19014501 1 g
02-190-14505 5 g
02-190-14510 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19014525 Supplier MP Biomedicals, Inc Supplier No. 0219014525
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26787-78-0
Molecular Formula C16H19N3O5S
Molecular Weight (g/mol) 365.404
InChI Key LSQZJLSUYDQPKJ-NJBDSQKTSA-N
Synonym Amoxycillin, Amolin, AMPC
PubChem CID 33613
ChEBI CHEBI:2676
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)O)C

Specifications

Type Amoxicillin
Physical Form Crystalline
Quantity 25 g
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Formula Weight 365.4
Percent Purity 95.0 to 102.0%
Grade Not Available
Chemical Name or Material Amoxicillin
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.