Learn More
Medchemexpress LLC Amoxicillin-d4 | 2673270-36-3 | 99.0% | 5 MG
SDP
Catalog No. 5000408530
Encompass_Preferred
Amoxicillin-d4 is a deuterium-labeled Amoxicillin, an antibiotic known for its good oral absorption and broad-spectrum antimicrobial activity. It serves as a tracer and an internal standard for quantitative analysis, particularly useful in methods like NMR, GC-MS, or LC-MS. Deuteration, which involves incorporating stable heavy isotopes into drug molecules, is used during drug development to track compounds and can influence pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Antibiotic with good oral absorption
  • Broad spectrum antimicrobial activity
Catalog No. 50-004-08530 Supplier Medchemexpress LLC Supplier No. HYB0467S5MG
Amoxicillin-d4 is a deuterium-labeled Amoxicillin, an antibiotic known for its good oral absorption and broad-spectrum antimicrobial activity. It serves as a tracer and an internal standard for quantitative analysis, particularly useful in methods like NMR, GC-MS, or LC-MS. Deuteration, which involves incorporating stable heavy isotopes into drug molecules, is used during drug development to track compounds and can influence pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Antibiotic with good oral absorption
  • Broad spectrum antimicrobial activity