Learn More
Medchemexpress LLC Amoxicillin-d4 | 2673270-36-3 | 99.7% | 1 MG
SDP
Catalog No. 5000408508
Encompass_Preferred
Amoxicillin-d4 is a deuterium labeled Amoxicillin, an antibiotic known for its good oral absorption and broad-spectrum antimicrobial activity. The incorporation of stable heavy isotopes of hydrogen can influence the pharmacokinetic and metabolic profiles of drugs, making this compound useful in drug development for quantitation.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-08508 Supplier Medchemexpress LLC Supplier No. HYB0467S1MG
Amoxicillin-d4 is a deuterium labeled Amoxicillin, an antibiotic known for its good oral absorption and broad-spectrum antimicrobial activity. The incorporation of stable heavy isotopes of hydrogen can influence the pharmacokinetic and metabolic profiles of drugs, making this compound useful in drug development for quantitation.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.