Learn More
Apexbio Technology LLC Amoxicillin Sodium 34642-77-8 10g
SDP
Catalog No. 5000092455
Encompass_Preferred
Amoxicillin Sodium (CAS 34642-77-8) is a small-molecule inhibitor targeting bacterial cell wall biosynthesis It is designed to disrupt peptidoglycan formation thereby compromising bacterial cell wall integrity Amoxicillin Sodium exerts its biological activity primarily through inhibition of peptidoglycan biosynthesis leading to bacterial lysis and cell death In in vitro studies Amoxicillin Sodium demonstrates broad-spectrum antibacterial inhibitory activity with IC50 values ranging from 0 1 g/mL to 10 g/mL depending on bacterial strain variability and experimental conditions Based on these pharmacological properties Amoxicillin Sodium holds research potential in bacterial physiology characterization antibiotic resistance evaluations and in vitro infection modeling studies
Catalog No. 50-000-92455 Supplier Apexbio Technology LLC Supplier No. B213710000
Amoxicillin Sodium (CAS 34642-77-8) is a small-molecule inhibitor targeting bacterial cell wall biosynthesis It is designed to disrupt peptidoglycan formation thereby compromising bacterial cell wall integrity Amoxicillin Sodium exerts its biological activity primarily through inhibition of peptidoglycan biosynthesis leading to bacterial lysis and cell death In in vitro studies Amoxicillin Sodium demonstrates broad-spectrum antibacterial inhibitory activity with IC50 values ranging from 0 1 g/mL to 10 g/mL depending on bacterial strain variability and experimental conditions Based on these pharmacological properties Amoxicillin Sodium holds research potential in bacterial physiology characterization antibiotic resistance evaluations and in vitro infection modeling studies