Learn More
Medchemexpress LLC Amoxicillin sodium | 34642-77-8 | 98.0% | 500 MG
SDP
Catalog No. 5000407850
Encompass_Preferred
Amoxicillin sodium is an antibiotic that exhibits good oral absorption and broad-spectrum antimicrobial activity. It functions by inhibiting the biosynthesis of polypeptides in the cell wall, which in turn inhibits cell growth.
  • Good oral absorption
  • Broad-spectrum antimicrobial activity
  • Inhibits the biosynthesis of polypeptides in the cell wall
  • Decreases living cells and increases the degree of cell wall rupture in a dose-dependent manner
  • Inhibits bacterial strain numbers and improves the survival rate in rats
  • Effective against infection with Chlamydia trachomatis in mice
Catalog No. 50-004-07850 Supplier Medchemexpress LLC Supplier No. HYB0467500MG
Amoxicillin sodium is an antibiotic that exhibits good oral absorption and broad-spectrum antimicrobial activity. It functions by inhibiting the biosynthesis of polypeptides in the cell wall, which in turn inhibits cell growth.
  • Good oral absorption
  • Broad-spectrum antimicrobial activity
  • Inhibits the biosynthesis of polypeptides in the cell wall
  • Decreases living cells and increases the degree of cell wall rupture in a dose-dependent manner
  • Inhibits bacterial strain numbers and improves the survival rate in rats
  • Effective against infection with Chlamydia trachomatis in mice