Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amoxicillin Sodium Salt, G-Biosciences
SCI_ED

Catalog No. S157222
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S157222 5 g
S000023 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S157222 Supplier G-Biosciences Supplier No. RC882
Add to Cart
Add to Cart

Amoxicillin is an extended-spectrum Beta-lactam antibiotic with a structure similar to that of ampicillin.

It is effective against a variety of Beta-lactamase negative bacteria including E. coli and various Streptococcus and Staphylococcus bacteria. It is also effective against a variety of other gram-negative and gram-positive bacteria.

Specifications

Chemical Name or Material Amoxicillin Sodium
CAS 34642-77-8
Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Industry Type Biotech
Molecular Weight (g/mol) 387.39 g/mol
Quantity 5 g
Percent Purity 97%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.