Learn More
Research Products International Corp Amoxicillin Trihydrate, 10 Grams
SDP
Catalog No. 50213134
Encompass_Preferred
Amoxicillin is a broad-spectrum, semi-synthetic penicillin antibiotic that is effective against a wide range of bacteria. The trihydrate form of amoxicillin is a crystalline powder that is often used in laboratory settings.  
Amoxicillin inhibits the growth of bacteria by interfering with their cell wall synthesis. It is a versatile antibiotic with low toxicity to mammalian cells, making it a valuable tool in maintaining sterile conditions in cell cultures and studying bacterial physiology.
Researchers use amoxicillin to selectively culture bacteria or to treat bacterial infections in cell cultures.
Catalog No. 50-213-134 Supplier Research Products International Corp Supplier No. A4002010.0
Amoxicillin is a broad-spectrum, semi-synthetic penicillin antibiotic that is effective against a wide range of bacteria. The trihydrate form of amoxicillin is a crystalline powder that is often used in laboratory settings.  
Amoxicillin inhibits the growth of bacteria by interfering with their cell wall synthesis. It is a versatile antibiotic with low toxicity to mammalian cells, making it a valuable tool in maintaining sterile conditions in cell cultures and studying bacterial physiology.
Researchers use amoxicillin to selectively culture bacteria or to treat bacterial infections in cell cultures.