Learn More
Selleck Chemical LLC Amoxicillin trihydrate S5298-25mg
SDP
Catalog No. 503112300
Encompass_Preferred
Amoxicillin (Amoxil Amoxipen Moxaline) is a broad-spectrum semisynthetic aminopenicillin antibiotic with bactericidal activity
Catalog No. 50-311-2300 Supplier Selleck Chemical LLC Supplier No. S529825MG
Amoxicillin (Amoxil Amoxipen Moxaline) is a broad-spectrum semisynthetic aminopenicillin antibiotic with bactericidal activity