Learn More
Apexbio Technology LLC Amoxicillin trihydrate 61336-70-7 1g
SDP
Catalog No. 502240555
Encompass_Preferred
Amoxicillin trihydrate (CAS 61336-70-7) is a small-molecule inhibitor targeting bacterial cell wall synthesis It is designed to inhibit peptidoglycan cross-linking thereby disrupting bacterial cell wall biosynthesis and resulting in cell lysis Amoxicillin trihydrate exerts its biological activity primarily through inhibition of bacterial transpeptidase enzymes involved in cell wall synthesis In in vitro studies Amoxicillin trihydrate demonstrates bactericidal activity with reported minimum inhibitory concentration (MIC) values of approximately 0 01 g/ml for Streptococcus viridans 0 25 g/ml for Bacillus anthracis and 50 g/ml for Retterella indica Based on these pharmacological properties Amoxicillin trihydrate holds research potential in antibacterial susceptibility testing microbial growth inhibition studies and in vitro pharmacological investigations
This item is not returnable. View return policy
Catalog No. 50-224-0555 Supplier Apexbio Technology LLC Supplier No. B33611000
Amoxicillin trihydrate (CAS 61336-70-7) is a small-molecule inhibitor targeting bacterial cell wall synthesis It is designed to inhibit peptidoglycan cross-linking thereby disrupting bacterial cell wall biosynthesis and resulting in cell lysis Amoxicillin trihydrate exerts its biological activity primarily through inhibition of bacterial transpeptidase enzymes involved in cell wall synthesis In in vitro studies Amoxicillin trihydrate demonstrates bactericidal activity with reported minimum inhibitory concentration (MIC) values of approximately 0 01 g/ml for Streptococcus viridans 0 25 g/ml for Bacillus anthracis and 50 g/ml for Retterella indica Based on these pharmacological properties Amoxicillin trihydrate holds research potential in antibacterial susceptibility testing microbial growth inhibition studies and in vitro pharmacological investigations