Learn More
Medchemexpress LLC Amoxicillin trihydrate | 61336-70-7 | 99.8% | 1 ML
SDP
Catalog No. 5000406771
Encompass_Preferred
Amoxicillin trihydrate is an antibiotic exhibiting good oral absorption and broad-spectrum antimicrobial activity. It functions by inhibiting the biosynthesis of polypeptides in the bacterial cell wall, thereby inhibiting cell growth.
  • Good oral absorption
  • Broad-spectrum antimicrobial activity
  • Inhibits biosynthesis of polypeptides in the cell wall
  • Inhibits cell growth
  • Suitable for in vitro studies
  • Effective in in vivo models against various infections
Catalog No. 50-004-06771 Supplier Medchemexpress LLC Supplier No. HYB0467B10MM/1ML
Amoxicillin trihydrate is an antibiotic exhibiting good oral absorption and broad-spectrum antimicrobial activity. It functions by inhibiting the biosynthesis of polypeptides in the bacterial cell wall, thereby inhibiting cell growth.
  • Good oral absorption
  • Broad-spectrum antimicrobial activity
  • Inhibits biosynthesis of polypeptides in the cell wall
  • Inhibits cell growth
  • Suitable for in vitro studies
  • Effective in in vivo models against various infections