Learn More
Indofine Chemical Amoxycillin Trihydrat, 10 Gm
SDP
Catalog No. 50910507
Encompass_Preferred
AMOXYCILLIN TRIHYDRATE 10 GM, Products for Life Science, Amoxicillin trihydrate, C16H19N3O5S 3 H2O, MW: 419.45, 61336-70-7, MFCD00072029
Catalog No. 50-910-507 Supplier Indofine Chemical Supplier No. BIO032 10 GM
AMOXYCILLIN TRIHYDRATE 10 GM, Products for Life Science, Amoxicillin trihydrate, C16H19N3O5S 3 H2O, MW: 419.45, 61336-70-7, MFCD00072029