Learn More
Enzo Life Sciences Adenosine 5'-monophosphate. disodium salt (5g). CAS: 4578-31-8
SDP
Catalog No. 502009304
Encompass_Preferred
Alternative name: AMP . 2Na. Purity: ≥98% (HPLC). Solubility: Soluble in water or aqueous buffers. Long Term Storage: -20°C.
Catalog No. 50-200-9304 Supplier Enzo Life Sciences Supplier No. ALX480014G005
Alternative name: AMP . 2Na. Purity: ≥98% (HPLC). Solubility: Soluble in water or aqueous buffers. Long Term Storage: -20°C.