Learn More
Medchemexpress LLC AMP-activated protein kinase gamma-1/beta-2/alpha-1 heterotrimer, human recombinant (His-GST) | 96.3% | 5 UG
SDP
Catalog No. 5000294222
Encompass_Preferred
Recombinant human AMPK heterotrimer containing γ1, β2, and α1 subunits, expressed in Sf9 insect cells and affinity-tagged with His-GST. This purified complex is provided for biochemical and cell-free assays requiring the intact AMPK enzyme complex.
  • Purity 96.3% by supplier analysis.
  • Expressed in Sf9 insect cells (baculovirus expression).
  • His-GST tag enables affinity purification and detection.
  • Approximate subunit molecular weights: ~35 kDa, ~37 kDa, ~95 kDa.
  • Supplied as a 5 μg quantity suitable for assay development.
  • Intended for biochemical and in vitro functional assays.
Catalog No. 50-002-94222 Supplier Medchemexpress LLC Supplier No. HYP761435UG
Recombinant human AMPK heterotrimer containing γ1, β2, and α1 subunits, expressed in Sf9 insect cells and affinity-tagged with His-GST. This purified complex is provided for biochemical and cell-free assays requiring the intact AMPK enzyme complex.
  • Purity 96.3% by supplier analysis.
  • Expressed in Sf9 insect cells (baculovirus expression).
  • His-GST tag enables affinity purification and detection.
  • Approximate subunit molecular weights: ~35 kDa, ~37 kDa, ~95 kDa.
  • Supplied as a 5 μg quantity suitable for assay development.
  • Intended for biochemical and in vitro functional assays.