Learn More
Medchemexpress LLC AMP-Deoxynojirimycin | 1MG
SDP
Catalog No. 5000230066
Encompass_Preferred
AMP-Deoxynojirimycin (AMP-DNM) is a potent ceramide glucosyltransferase and GCase 2 inhibitor AMP-Deoxynojirimycin also is a GlcCer biosynthesis inhibitor[1][2]
Catalog No. 50-002-30066 Supplier Medchemexpress LLC Supplier No. HY1146151MG
AMP-Deoxynojirimycin (AMP-DNM) is a potent ceramide glucosyltransferase and GCase 2 inhibitor AMP-Deoxynojirimycin also is a GlcCer biosynthesis inhibitor[1][2]