Learn More
Electron Microscopy Sciences AMP Passive Air Isolation Platform with White Epoxy Surface, 90 x 150 cm
SDP
Catalog No. 5034454
Encompass_Preferred
These High Performance anti-vibration platforms for microscopes are designed to be pumped-up and left alone. Simple to use, they are ideal for use with fixed loads or in applications where pressurised air or gas cannot be used. The unique air damped rolling diaphragm sub 2Hz frequency isolator removes up to 99 percent of vibrations. And sound deadening construction eliminates unwanted surface vibrations. They support loads up to 100kg .
Size: 90 x 150 cm
Surface: White Epoxy
Catalog No. 50-344-54 Supplier Electron Microscopy Sciences Supplier No. 6410
These High Performance anti-vibration platforms for microscopes are designed to be pumped-up and left alone. Simple to use, they are ideal for use with fixed loads or in applications where pressurised air or gas cannot be used. The unique air damped rolling diaphragm sub 2Hz frequency isolator removes up to 99 percent of vibrations. And sound deadening construction eliminates unwanted surface vibrations. They support loads up to 100kg .
Size: 90 x 150 cm
Surface: White Epoxy