Learn More
Medchemexpress LLC AMP-PCP disodium 5mg | 7414-56-4 | 5MG
SDP
Catalog No. 5000240106
Encompass_Preferred
AMP-PCP disodium is an ATP analogue and can bind to Hsp90 N-terminal domain with a Kd value of 3 8 M AMP-PCP disodium binding favors the formation of the active homodimer of Hsp90[1
Catalog No. 50-002-40106 Supplier Medchemexpress LLC Supplier No. HY106723A5MG
AMP-PCP disodium is an ATP analogue and can bind to Hsp90 N-terminal domain with a Kd value of 3 8 M AMP-PCP disodium binding favors the formation of the active homodimer of Hsp90[1