Learn More
TARGETMOL CHEMICALS INC AMP-PCP disodium 5MG
SDP
Catalog No. 502879600
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. AMP-PCP disodium, an ATP analogue, binds to the N-terminal domain of Hsp90 with a Kd value of 3.8 uM, facilitating the formation of an active Hsp90 homodimer. Purity 100%
Catalog No. 50-287-9600 Supplier TARGETMOL CHEMICALS INC Supplier No. T13547L5MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. AMP-PCP disodium, an ATP analogue, binds to the N-terminal domain of Hsp90 with a Kd value of 3.8 uM, facilitating the formation of an active Hsp90 homodimer. Purity 100%