Learn More
Bioworld AMP SEPAROPOREAGAROSE) 4B
Supplier:  Bioworld 201810792
Encompass
AMP SEPAROPORE (AGAROSE) 4B-CL, LYOPHILIZED: AMP Separopore The method involves alkylation of the parent nucleotide, AMP followed by alkaline rearrangement to yield the corresponding N6- carboxymethyl derivative with ubsequent condensat...
Catalog No. 50-197-874