Learn More
Enzo Life Sciences AMPA (5mg). CAS: 77521-29-0
SDP
Supplier:  Enzo Life Sciences BMLEA1100005
Encompass_Preferred
Potent and selective quisqualate analog that interacts with the AMPA subgroup of ionotropic glutamate receptors. Purity: ≥98% (TLC). Solubility: Soluble in water (2mg/ml, warm) or DMSO (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0523