Learn More
Apexbio Technology LLC Ampalex 154235-83-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000093387
Encompass_Preferred
Ampalex (Ampakine CX516 CAS 154235-83-3) is a small molecule modulator that selectively enhances the activity of AMPA-type glutamate receptors through positive allosteric modulation By facilitating glutamatergic neurotransmission Ampalex has been shown to improve functional deficits in AMPA receptors within hippocampal slice models exhibiting protein accumulation This mechanism underpins its exploration as a research tool in studies of neurodegenerative and neuropsychiatric disorders including Alzheimer s disease schizophrenia and mild cognitive impairment Ampalex is utilized to investigate the therapeutic potential of targeting AMPA receptors in these disease states
Catalog No. 50-000-93387 Supplier Apexbio Technology LLC Supplier No. A31765.1
Ampalex (Ampakine CX516 CAS 154235-83-3) is a small molecule modulator that selectively enhances the activity of AMPA-type glutamate receptors through positive allosteric modulation By facilitating glutamatergic neurotransmission Ampalex has been shown to improve functional deficits in AMPA receptors within hippocampal slice models exhibiting protein accumulation This mechanism underpins its exploration as a research tool in studies of neurodegenerative and neuropsychiatric disorders including Alzheimer s disease schizophrenia and mild cognitive impairment Ampalex is utilized to investigate the therapeutic potential of targeting AMPA receptors in these disease states