Learn More
Ampco Safety Tools Socket Wrenches
SDP
Catalog No. 19166481 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
No. per Pack:
10 Piece Set
21 Piece Set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
19-166-481 21 Piece Set
19-166-482 10 Piece Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-166-481 Supplier Ampco Safety Tools Supplier No. W260
Add to Cart
Add to Cart
Specifications
Product Type Square Drive Socket Wrench
Drive Size 0.5 in.
Material Aluminum Bronze
Includes 0.31 to 1.25 in. (7) Sockets, ratchet, sliding bar, extension bar, hinged handle and case
No. per Pack 21 Piece Set
Weight (English) 13.3 lb.