Learn More
Ampco Safety Tools Socket Wrenches
SDP
Catalog No. 19166482 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
No. per Pack:
10 Piece Set
21 Piece Set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
19-166-482 10 Piece Set
19-166-481 21 Piece Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-166-482 Supplier Ampco Safety Tools Supplier No. W290
Specifications
Product Type Square Drive Socket Wrench
Drive Size 0.5 in.
Material Aluminum Bronze
Includes 5 Sockets, ratchet, sliding bar, extension bar, hinged handle and case
No. per Pack 10 Piece Set
Weight (English) 10.2 lb.