Learn More
Enzo Life Sciences AMP'D HSP70 high sensitivity ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ENZKIT1010001
Encompass_Preferred
The AMP'D HSP70 high sensitivity ELISA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4.5 hours. Alternative name: Heat shock protein 70, HSP72. Assay Time: 4.5 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human, Mouse, Rat .
Catalog No. 50-201-3805