Learn More
Aviva Systems Biology Corp AMPD2 Antibody - N-terminal region (ARP64642_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64642_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AMPD2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human AMPD2. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-322