Learn More
Aviva Systems Biology Corp AMPD3 Antibody - C-terminal region (ARP64725_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64725_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AMPD3. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-288-345