Learn More
SMITH ECOLOGICAL SYSTEMS AMPEROMETRIC TITRATOR
Catalog No. NC9242374
Encompass
Amperometric Titrator, Switch Range: Std. Down to 0.01 ppm, Front Titration Procedure, Back Titration, Highly Sensitive Meter, Adjustable Potentiometer, Sulfur Dioxide, Ideal for Waste Water, Dimension: 24 in. H x 8- 5/8 in. W x 7- 1/2
Catalog No. NC9242374 Supplier SMITH ECOLOGICAL SYSTEMS Supplier No. W3T108068 / A790
Add to Cart
Add to Cart
Request a Quote
Amperometric Titrator, Switch Range: Std. Down to 0.01 ppm, Front Titration Procedure, Back Titration, Highly Sensitive Meter, Adjustable Potentiometer, Sulfur Dioxide, Ideal for Waste Water, Dimension: 24 in. H x 8- 5/8 in. W x 7- 1/2