Learn More
Medchemexpress LLC Amperozide | 75558-90-6 | >98.0% | 5 MG

Supplier: Medchemexpress LLC HY1213015MG
Amperozide is a diphenylbutylpiperazine atypical antipsychotic research compound that functions primarily as a 5-HT2 receptor antagonist (Ki ≈ 26 nM) and exhibits low affinity for D2 receptors. It is supplied as a high-purity reagent for laboratory research.
- CAS number: 75558-90-6
- Molecular formula: C23H29F2N3O
- Molecular weight: 401.5 g/mol
- Purity: >98%
- Typical pack size: 5 mg
- Primary activity: 5-HT2 receptor antagonist, low D2 affinity
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.