Learn More
Electron Microscopy Sciences Amphibian Ringer Solution 500 ML
SDP
Catalog No. 50980243
Encompass_Preferred
Amphibian Ringer Solution is made up of salts including sodium chloride, potassium chloride, and calcium chloride hexahydrate.
Catalog No. 50-980-243 Supplier Electron Microscopy Sciences Supplier No. 1176205
Amphibian Ringer Solution is made up of salts including sodium chloride, potassium chloride, and calcium chloride hexahydrate.