Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amphiregulin Human 10ug
SDP

Catalog No. 5000184403
Encompass_Preferred

Recombinant human amphiregulin is a tag-free, lyophilized epidermal growth factor receptor (EGFR) ligand that promotes proliferation of EGFR-responsive cells. It is supplied with high purity and low endotoxin and is intended for use in cell-based proliferation assays and biochemical studies; follow the manufacturer's reconstitution and storage recommendations for best results.

  • Tag free, lyophilized powder for convenient storage and handling.
  • High purity (>95% by reducing SDS-PAGE) for consistent experimental results.
  • Low endotoxin (<1 EU/μg) suitable for cell-based assays.
  • Demonstrated biological activity with ED50 ≤124.1 ng/mL in a cell proliferation assay.
  • Recommended storage at -20°C and aliquot freezing for long-term stability.

Catalog No. 50-001-84403 Supplier Medchemexpress LLC Supplier No. HYP7057810UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human amphiregulin is a tag-free, lyophilized epidermal growth factor receptor (EGFR) ligand that promotes proliferation of EGFR-responsive cells. It is supplied with high purity and low endotoxin and is intended for use in cell-based proliferation assays and biochemical studies; follow the manufacturer's reconstitution and storage recommendations for best results.

  • Tag free, lyophilized powder for convenient storage and handling.
  • High purity (>95% by reducing SDS-PAGE) for consistent experimental results.
  • Low endotoxin (<1 EU/μg) suitable for cell-based assays.
  • Demonstrated biological activity with ED50 ≤124.1 ng/mL in a cell proliferation assay.
  • Recommended storage at -20°C and aliquot freezing for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.