Learn More
Genscript Corporation Antibody Protein Proteins Proteins 50 UG 1/EA
Supplier:  Genscript Corporation Z0310350
Encompass
Amphiregulin is a member of the EGF family of cytokines which comprises at least ten proteins including EGF TGF-Alpha HB-EGF Epiregulin Tomoregulin Neuregulins and the various heregulins. Through the EGF/TGF-Alpha receptor it stimulates growth of keratinocytes epithelial cells and some fibroblasts. Amphiregulin also inhibits the growth of certain carcinoma cell lines. Synthesized as a transmembrane protein Amphiregulins extracellular domain is proteolytically processed to release the mature protein.
Catalog No. 50-252-9838