Learn More
Medchemexpress LLC Amphiregulin Mouse 20ug | 20ug
SDP
Catalog No. 5000193612
Encompass_Preferred
Amphiregulin Protein an EGFR ligand acts as an autocrine growth factor and mitogen for diverse cells including astrocytes Schwann cells and fibroblasts Engaging with CNIH in its immature stage Amphiregulin s EGFR activation emphasizes its vital role in regulating cell growth and orchestrating various cellular functions across different cell types Amphiregulin Protein Mouse (HEK293 Fc) is the recombinant mouse-derived Amphiregulin protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-93612 Supplier Medchemexpress LLC Supplier No. HYP7786820UG
Amphiregulin Protein an EGFR ligand acts as an autocrine growth factor and mitogen for diverse cells including astrocytes Schwann cells and fibroblasts Engaging with CNIH in its immature stage Amphiregulin s EGFR activation emphasizes its vital role in regulating cell growth and orchestrating various cellular functions across different cell types Amphiregulin Protein Mouse (HEK293 Fc) is the recombinant mouse-derived Amphiregulin protein expressed by HEK293 with N-hFc labeled tag