Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Amphotericin B/Penicillin/Streptomycin, 50 mL
SDP

Catalog No. 507517247
Encompass_Preferred

Amphotericin B, Penicillin, Streptomycin Mixture is useful for reducing the chance of microbial contamination.

100X concentration - dilute 1:100 in cell culture medium

Amphotericin B: 25 µg/mL
Penicillin: 10,000 Units/mL
Streptomycin: 10,000 µg/mL

Storage Condition: -10 °C to -20 °C
Shipping Condition: on dry ice
Sterile Filtered
Cytoxity Tested
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7247 Supplier Quality Biological Inc Supplier No. 120096711EA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amphotericin B, Penicillin, Streptomycin Mixture is useful for reducing the chance of microbial contamination.

100X concentration - dilute 1:100 in cell culture medium

Amphotericin B: 25 µg/mL
Penicillin: 10,000 Units/mL
Streptomycin: 10,000 µg/mL

Storage Condition: -10 °C to -20 °C
Shipping Condition: on dry ice
Sterile Filtered
Cytoxity Tested
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.