Learn More
Alkali Scientific Amphotericin B, 100 G
SDP
Catalog No. 5000660731
Encompass_Preferred
Amphotericin B is a highly effective antifungal compound used extensively in laboratory and research applications to control fungal contamination. It acts by binding to ergosterol within fungal cell membranes, disrupting membrane permeability and leading to cell death. This makes it particularly useful for inhibiting a wide range of yeasts and molds in culture systems. In microbiology and cell culture laboratories, it is commonly added to media to maintain sterile conditions during long incubation periods. Amphotericin B is also used in antifungal research and drug development studies due to its strong and well documented antifungal properties.
Catalog No. 50-006-60731 Supplier Alkali Scientific Supplier No. AMP004CP
Amphotericin B is a highly effective antifungal compound used extensively in laboratory and research applications to control fungal contamination. It acts by binding to ergosterol within fungal cell membranes, disrupting membrane permeability and leading to cell death. This makes it particularly useful for inhibiting a wide range of yeasts and molds in culture systems. In microbiology and cell culture laboratories, it is commonly added to media to maintain sterile conditions during long incubation periods. Amphotericin B is also used in antifungal research and drug development studies due to its strong and well documented antifungal properties.