Learn More
Alkali Scientific Amphotericin B, 100 Mg
SDP
Catalog No. 5000660435
Encompass_Preferred
Amphotericin B is a polyene antifungal agent widely used in microbiology and cell culture applications to prevent and control fungal contamination. It functions by binding to ergosterol in fungal cell membranes, creating pores that disrupt membrane integrity and lead to cell death. This mechanism makes it highly effective against a broad range of yeasts and molds. In laboratory settings, Amphotericin B is commonly added to culture media to maintain sterile conditions during experiments. It is also used in research involving fungal pathogens, antifungal susceptibility testing, and studies focused on membrane biology and antifungal drug development.
Catalog No. 50-006-60435 Supplier Alkali Scientific Supplier No. AMP001CP
Amphotericin B is a polyene antifungal agent widely used in microbiology and cell culture applications to prevent and control fungal contamination. It functions by binding to ergosterol in fungal cell membranes, creating pores that disrupt membrane integrity and lead to cell death. This mechanism makes it highly effective against a broad range of yeasts and molds. In laboratory settings, Amphotericin B is commonly added to culture media to maintain sterile conditions during experiments. It is also used in research involving fungal pathogens, antifungal susceptibility testing, and studies focused on membrane biology and antifungal drug development.