Learn More
Crescent Chemical Co Inc AMPHOTERICIN B 100 MG
SDP
Catalog No. 5072528
Encompass_Preferred
Amphotericin B Pharma./veter. comp. (WS) CAS 1397-89-3 Molecular Weight - 924.079 Storage - -18°C Catalog Num - DRE-C10243050 chemical
Catalog No. 50-725-28 Supplier Crescent Chemical Co Inc Supplier No. PSC10243050
Amphotericin B Pharma./veter. comp. (WS) CAS 1397-89-3 Molecular Weight - 924.079 Storage - -18°C Catalog Num - DRE-C10243050 chemical