Learn More
Medchemexpress LLC Amphotericin B | 1397-89-3 | 93.3% | 1 G
SDP
Catalog No. 5000405640
Encompass_Preferred
Amphotericin B is a polyene antifungal agent effective against a wide range of fungal pathogens. It works by irreversibly binding to ergosterol, which leads to the disruption of membrane integrity and ultimately causes cell death. This product is for research use only.
  • Polyene antifungal agent
  • Effective against a wide range of fungal pathogens
  • Binds irreversibly to ergosterol
  • Disrupts membrane integrity, causing cell death
  • Intended for research use only
Catalog No. 50-004-05640 Supplier Medchemexpress LLC Supplier No. HYB02211G
Amphotericin B is a polyene antifungal agent effective against a wide range of fungal pathogens. It works by irreversibly binding to ergosterol, which leads to the disruption of membrane integrity and ultimately causes cell death. This product is for research use only.
  • Polyene antifungal agent
  • Effective against a wide range of fungal pathogens
  • Binds irreversibly to ergosterol
  • Disrupts membrane integrity, causing cell death
  • Intended for research use only