Learn More
Medchemexpress LLC Amphotericin B | 1397-89-3 | 93.3% | 1 ML
SDP
Catalog No. 5000407188
Encompass_Preferred
Amphotericin B is a polyene antifungal agent effective against a wide range of fungal pathogens. Its mechanism of action involves irreversibly binding to ergosterol, which leads to the disruption of the fungal cell membrane integrity and ultimately cell death.
  • Antifungal agent
  • Effective against a wide range of fungal pathogens
  • Disrupts fungal cell membrane integrity
  • Leads to cell death
Catalog No. 50-004-07188 Supplier Medchemexpress LLC Supplier No. HYB022110MM/1ML
Amphotericin B is a polyene antifungal agent effective against a wide range of fungal pathogens. Its mechanism of action involves irreversibly binding to ergosterol, which leads to the disruption of the fungal cell membrane integrity and ultimately cell death.
  • Antifungal agent
  • Effective against a wide range of fungal pathogens
  • Disrupts fungal cell membrane integrity
  • Leads to cell death