Learn More
Medchemexpress LLC Amphotericin B | 1397-89-3 | 93.3% | 500 MG
SDP
Catalog No. 5000405718
Encompass_Preferred
Amphotericin B is a polyene antifungal agent effective against a wide range of fungal pathogens. It functions by irreversibly binding to ergosterol, which leads to the disruption of the fungal cell membrane integrity and ultimately results in cell death. This product is for research use only.
  • Polyene antifungal agent
  • Effective against a wide range of fungal pathogens
  • Disrupts fungal cell membrane integrity
  • Leads to cell death
Catalog No. 50-004-05718 Supplier Medchemexpress LLC Supplier No. HYB0221500MG
Amphotericin B is a polyene antifungal agent effective against a wide range of fungal pathogens. It functions by irreversibly binding to ergosterol, which leads to the disruption of the fungal cell membrane integrity and ultimately results in cell death. This product is for research use only.
  • Polyene antifungal agent
  • Effective against a wide range of fungal pathogens
  • Disrupts fungal cell membrane integrity
  • Leads to cell death