Learn More
Apexbio Technology LLC Amphotericin B 1397-89-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062281
Encompass_Preferred
Amphotericin B is a polyene antifungal antibiotic derived from Streptomyces nodosus It exerts its antifungal action primarily by binding sterols in fungal membranes forming pores that disrupt membrane integrity This binding induces altered ion permeability permitting movement of small ions including potassium and sodium across the membrane Additionally Amphotericin B stimulates inflammatory cytokine release in immune cells through interaction with Toll-like receptor 2 (TLR2) and CD14-dependent pathways Amphotericin B is used in biomedical research to investigate fungal infection mechanisms immune signaling events mediated by TLR pathways and prion-related disorders Its antifungal activity exhibits an IC50 range from 0 028 to 0 290 g/ml in vitro
Catalog No. 50-000-62281 Supplier Apexbio Technology LLC Supplier No. B18855.1
Amphotericin B is a polyene antifungal antibiotic derived from Streptomyces nodosus It exerts its antifungal action primarily by binding sterols in fungal membranes forming pores that disrupt membrane integrity This binding induces altered ion permeability permitting movement of small ions including potassium and sodium across the membrane Additionally Amphotericin B stimulates inflammatory cytokine release in immune cells through interaction with Toll-like receptor 2 (TLR2) and CD14-dependent pathways Amphotericin B is used in biomedical research to investigate fungal infection mechanisms immune signaling events mediated by TLR pathways and prion-related disorders Its antifungal activity exhibits an IC50 range from 0 028 to 0 290 g/ml in vitro