Learn More
Alkali Scientific Amphotericin B 250µg/mL Solution, 50 Milliliters
SDP
Catalog No. 5000660127
Encompass_Preferred
Amphotericin B solution is a ready to use antifungal preparation formulated for use in cell culture and microbiological applications. It functions by binding to ergosterol in fungal cell membranes, causing structural disruption and leakage of intracellular components, ultimately leading to fungal cell death. The pre prepared solution format allows easy and accurate addition to culture systems without the need for dissolution or preparation steps. It is commonly used to prevent fungal contamination in cell cultures, tissue cultures, and laboratory experiments. This solution is also suitable for antifungal research and studies requiring controlled inhibition of fungal growth.
Catalog No. 50-006-60127 Supplier Alkali Scientific Supplier No. MPH001CP
Amphotericin B solution is a ready to use antifungal preparation formulated for use in cell culture and microbiological applications. It functions by binding to ergosterol in fungal cell membranes, causing structural disruption and leakage of intracellular components, ultimately leading to fungal cell death. The pre prepared solution format allows easy and accurate addition to culture systems without the need for dissolution or preparation steps. It is commonly used to prevent fungal contamination in cell cultures, tissue cultures, and laboratory experiments. This solution is also suitable for antifungal research and studies requiring controlled inhibition of fungal growth.