Learn More
Alkali Scientific Amphotericin B, 250 Mg
SDP
Catalog No. 5000660196
Encompass_Preferred
Amphotericin B is a potent antifungal compound used in laboratory research and cell culture applications to inhibit fungal growth. It targets ergosterol in fungal cell membranes, disrupting membrane structure and causing leakage of cellular contents. This action leads to effective elimination of yeasts and molds in contaminated samples. The compound is widely used in microbiology laboratories to maintain sterile culture conditions and prevent fungal overgrowth in experimental systems. Amphotericin B is also utilized in antifungal susceptibility studies and research focused on fungal physiology, providing reliable and well established antifungal activity.
Catalog No. 50-006-60196 Supplier Alkali Scientific Supplier No. AMP002CP
Amphotericin B is a potent antifungal compound used in laboratory research and cell culture applications to inhibit fungal growth. It targets ergosterol in fungal cell membranes, disrupting membrane structure and causing leakage of cellular contents. This action leads to effective elimination of yeasts and molds in contaminated samples. The compound is widely used in microbiology laboratories to maintain sterile culture conditions and prevent fungal overgrowth in experimental systems. Amphotericin B is also utilized in antifungal susceptibility studies and research focused on fungal physiology, providing reliable and well established antifungal activity.