Learn More
ILEX LIFE SCIENCES LLC AMPHOTERICIN B 250UG/ML 50ML
SDP
Catalog No. 502733747
Encompass_Preferred
PAN-Biotech Amphotericin B for cell culture (Fungizone generic), 250 ug/ml (50 ml). DESCRIPTION: PAN-Biotech Amphotericin B (250 ug/ml) is a generic version of Fungizone that is used in cell culture applications. Amphotericin B is an antifungal agent produced by Streptomyces sp.. Amphotericin B increases fungal plasma membrane permeability by binding to sterols, thereby protecting cell cultures from contamination by yeast and multicellular fungi. APPLICATION: Amphotericin B is often used as an antifungal, anti-yeast and anti-mold agent to prevent fungal contamination of cell cultures. The commonly recommended working concentration ranges from 0.25-2.5 ug/mL (1-10 mL/L). LIQUID/POWDER: Liquid. STERILE: Yes. STORAGE TEMP.: -20°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. USAGE: This product is intended for Laboratory Research Use Only. Not for use in diagnostic or therapeutic procedures.
Catalog No. 50-273-3747 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0601050
PAN-Biotech Amphotericin B for cell culture (Fungizone generic), 250 ug/ml (50 ml). DESCRIPTION: PAN-Biotech Amphotericin B (250 ug/ml) is a generic version of Fungizone that is used in cell culture applications. Amphotericin B is an antifungal agent produced by Streptomyces sp.. Amphotericin B increases fungal plasma membrane permeability by binding to sterols, thereby protecting cell cultures from contamination by yeast and multicellular fungi. APPLICATION: Amphotericin B is often used as an antifungal, anti-yeast and anti-mold agent to prevent fungal contamination of cell cultures. The commonly recommended working concentration ranges from 0.25-2.5 ug/mL (1-10 mL/L). LIQUID/POWDER: Liquid. STERILE: Yes. STORAGE TEMP.: -20°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. USAGE: This product is intended for Laboratory Research Use Only. Not for use in diagnostic or therapeutic procedures.