Learn More
Alkali Scientific Amphotericin B, 5 G
SDP
Catalog No. 5000660322
Encompass_Preferred
Amphotericin B is a polyene antifungal agent widely used in laboratory research to prevent fungal contamination in cell culture and microbiological studies. It interacts with ergosterol in fungal membranes, creating pores that disrupt membrane integrity and result in cell death. This makes it highly effective against a broad range of fungi. In laboratory workflows, it is commonly used to maintain sterile culture environments and protect experimental samples from fungal overgrowth. Amphotericin B is also employed in antifungal susceptibility testing and research focused on fungal biology, providing consistent and reliable antifungal activity.
Catalog No. 50-006-60322 Supplier Alkali Scientific Supplier No. AMP005CP
Amphotericin B is a polyene antifungal agent widely used in laboratory research to prevent fungal contamination in cell culture and microbiological studies. It interacts with ergosterol in fungal membranes, creating pores that disrupt membrane integrity and result in cell death. This makes it highly effective against a broad range of fungi. In laboratory workflows, it is commonly used to maintain sterile culture environments and protect experimental samples from fungal overgrowth. Amphotericin B is also employed in antifungal susceptibility testing and research focused on fungal biology, providing consistent and reliable antifungal activity.