Learn More
Apexbio Technology LLC Amphotericin B, 500mg Cas# 1397-89-3
SDP
Supplier:  Apexbio Technology LLC B1885500
Encompass_Preferred
Amphotericin B is a polyene, antifungal antibiotic produced from a strain of Streptomyces nodosus. (IC50=0.028-0.290 ug/ml) 1397-89-3. MFCD00877763 Amphotericin B, a polyene antifungal antibiotic, has been produced from a strain of Streptomyces nodosus with an IC50 of 0.0280.290 g/ml.In vitro: Amphotericin B was the most effective drug for treating many life-threatening fungal infections. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8656